siRNA Details
Gene Symbol: Hmgxb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107412.1 | NP_001100882.1 | 4 | 389 | ||
XM_006255141.2 | XP_006255203.1 | 3 | 4807 | |||
XM_006255143.1 | XP_006255205.1 | 4 | 391 | |||
XM_006255144.1 | XP_006255206.1 | 1 | 81 | |||
XM_008772312.1 | XP_008770534.1 | 1 | 259 |
