siRNA Details
| Gene Symbol: Hmgxb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107412.1 | NP_001100882.1 | 6 | 1385 | ||
| XM_006255141.2 | XP_006255203.1 | 5 | 5803 | |||
| XM_006255143.1 | XP_006255205.1 | 6 | 1387 | |||
| XM_006255144.1 | XP_006255206.1 | 4 | 1192 | |||
| XM_008772312.1 | XP_008770534.1 | 4 | 1370 | |||

Hmgxb4
Genomic Map