siRNA Details
| Gene Symbol: Irx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102413.1 | NP_001095883.1 | 5 | 1959 | ||
| XM_006255173.2 | XP_006255235.1 | 5 | 1938 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irx6| Gene Symbol: Irx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102413.1 | NP_001095883.1 | 5 | 1959 | ||
| XM_006255173.2 | XP_006255235.1 | 5 | 1938 | |||