siRNA Details
| Gene Symbol: Rpgrip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107414.1 | NP_001100884.1 | 4 | 341 | ||
| XM_008772313.1 | XP_008770535.1 | 3 | 769 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpgrip1l| Gene Symbol: Rpgrip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107414.1 | NP_001100884.1 | 4 | 341 | ||
| XM_008772313.1 | XP_008770535.1 | 3 | 769 | |||