siRNA Details
Gene Symbol: LOC307727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047882.1 | NP_001041347.1 | 1 | 239 | ||
GenBank mRNA | AY539920.1 | - | 1 | 239 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC307727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047882.1 | NP_001041347.1 | 1 | 239 | ||
GenBank mRNA | AY539920.1 | - | 1 | 239 |