siRNA Details
| Gene Symbol: LOC307727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047882.1 | NP_001041347.1 | 1 | 141 | ||
| GenBank mRNA | AY539920.1 | - | 1 | 141 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC307727| Gene Symbol: LOC307727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047882.1 | NP_001041347.1 | 1 | 141 | ||
| GenBank mRNA | AY539920.1 | - | 1 | 141 | ||