siRNA Details
| Gene Symbol: Gpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012057.1 | NP_001012057.1 | 4 | 563 | ||
| XM_008772405.1 | XP_008770627.1 | 5 | 684 | |||
| XM_008772406.1 | XP_008770628.1 | 5 | 873 | |||
| XM_008772407.1 | XP_008770629.1 | 5 | 610 | |||
| GenBank mRNA | AY325245.1 | - | 633 | |||
| FQ223845.1 | - | 759 | ||||

Gpt2
Genomic Map