siRNA Details
| Gene Symbol: Otud4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191700.1 | NP_001178629.1 | 21 | 3467 | ||
| XM_006255402.2 | XP_006255464.1 | 21 | 3806 | |||
| XM_008772493.1 | XP_008770715.1 | 21 | 3982 | |||
| XM_008772494.1 | XP_008770716.1 | 21 | 3538 | |||
| XM_008772495.1 | XP_008770717.1 | 22 | 3586 | |||
| GenBank mRNA | BC083886.1 | - | 4 | 1588 | ||
| BC100159.1 | - | 3 | 1372 | |||

Otud4
Genomic Map