siRNA Details
| Gene Symbol: Otud4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191700.1 | NP_001178629.1 | 21 | 3287 | ||
| XM_006255402.2 | XP_006255464.1 | 21 | 3626 | |||
| XM_008772493.1 | XP_008770715.1 | 21 | 3802 | |||
| XM_008772494.1 | XP_008770716.1 | 21 | 3358 | |||
| XM_008772495.1 | XP_008770717.1 | 22 | 3406 | |||
| GenBank mRNA | BC083886.1 | - | 4 | 1408 | ||
| BC100159.1 | - | 3 | 1192 | |||

Otud4
Genomic Map