siRNA Details
| Gene Symbol: Gfod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107421.1 | NP_001100891.1 | 3 | 957 | ||
| XM_008772497.1 | XP_008770719.1 | 3 | 2011 | |||
| XM_008772498.1 | XP_008770720.1 | 3 | 1551 | |||
| GenBank mRNA | BC161901.1 | - | 3 | 939 | 8367133 | |

Gfod2
Genomic Map