siRNA Details
| Gene Symbol: Slc7a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107424.1 | NP_001100894.1 | 7 | 1268 | ||
| XM_006255498.2 | XP_006255560.1 | 8 | 1333 | |||
| XM_006255499.2 | XP_006255561.1 | 8 | 1363 | |||
| XM_006255500.2 | XP_006255562.1 | 8 | 1481 | |||
| XM_008772500.1 | XP_008770722.1 | 8 | 1367 | |||
| XM_008772501.1 | XP_008770723.1 | 8 | 1548 | |||
| XM_008772502.1 | XP_008770724.1 | 8 | 1597 | |||

Slc7a6
Genomic Map