siRNA Details
| Gene Symbol: Slc7a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107424.1 | NP_001100894.1 | 6, 7 | 1149 | ||
| XM_006255498.2 | XP_006255560.1 | 7, 8 | 1214 | |||
| XM_006255499.2 | XP_006255561.1 | 7, 8 | 1244 | |||
| XM_006255500.2 | XP_006255562.1 | 7, 8 | 1362 | |||
| XM_008772500.1 | XP_008770722.1 | 7, 8 | 1248 | |||
| XM_008772501.1 | XP_008770723.1 | 7, 8 | 1429 | |||
| XM_008772502.1 | XP_008770724.1 | 7, 8 | 1478 | |||

Slc7a6
Genomic Map