siRNA Details
| Gene Symbol: Psmd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107426.1 | NP_001100896.1 | 4, 5 | 494 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmd7| Gene Symbol: Psmd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107426.1 | NP_001100896.1 | 4, 5 | 494 | ||