siRNA Details
| Gene Symbol: Chst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107427.1 | NP_001100897.1 | 2 | 150 | ||
| XM_006255586.2 | XP_006255648.1 | 2 | 149 | |||
| XM_006255587.2 | XP_006255649.1 | 2 | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chst4| Gene Symbol: Chst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107427.1 | NP_001100897.1 | 2 | 150 | ||
| XM_006255586.2 | XP_006255648.1 | 2 | 149 | |||
| XM_006255587.2 | XP_006255649.1 | 2 | 232 | |||