siRNA Details
Gene Symbol: Fuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107429.1 | NP_001100899.1 | 11 | 1039 | ||
XM_006255624.2 | XP_006255686.2 | 11 | 1051 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107429.1 | NP_001100899.1 | 11 | 1039 | ||
XM_006255624.2 | XP_006255686.2 | 11 | 1051 |