siRNA Details
| Gene Symbol: Fa2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135583.1 | NP_001129055.1 | 3 | 533 | ||
| GenBank mRNA | DQ339484.1 | - | 3 | 533 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fa2h| Gene Symbol: Fa2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135583.1 | NP_001129055.1 | 3 | 533 | ||
| GenBank mRNA | DQ339484.1 | - | 3 | 533 | ||