siRNA Details
| Gene Symbol: Ldhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008893.1 | NP_001008893.1 | 6 | 829 | ||
| XM_006255631.2 | XP_006255693.1 | 6 | 880 | |||
| GenBank mRNA | AY321341.1 | - | 6 | 829 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ldhd| Gene Symbol: Ldhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008893.1 | NP_001008893.1 | 6 | 829 | ||
| XM_006255631.2 | XP_006255693.1 | 6 | 880 | |||
| GenBank mRNA | AY321341.1 | - | 6 | 829 | ||