siRNA Details
| Gene Symbol: Chst5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107431.1 | NP_001100901.1 | 2 | 1016 | ||
| XM_006255632.2 | XP_006255694.1 | 3 | 1160 | |||
| XM_006255633.2 | XP_006255695.1 | 3 | 1794 | |||
| GenBank mRNA | FQ212819.1 | - | 3 | 1160 | ||
| FQ213398.1 | - | 2 | 1050 | |||

Chst5
Genomic Map