siRNA Details
| Gene Symbol: Terf2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013143.1 | NP_001013161.1 | 3 | 1156 | ||
| GenBank mRNA | BC090335.1 | - | 3 | 1156 | 7302641 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Terf2ip| Gene Symbol: Terf2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013143.1 | NP_001013161.1 | 3 | 1156 | ||
| GenBank mRNA | BC090335.1 | - | 3 | 1156 | 7302641 | |