siRNA Details
| Gene Symbol: Cntnap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107432.1 | NP_001100902.1 | 18 | 3296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntnap4| Gene Symbol: Cntnap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107432.1 | NP_001100902.1 | 18 | 3296 | ||