siRNA Details
| Gene Symbol: FZD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003507.1 | NP_003498.1 | 1 | 1400 | ||
| GenBank mRNA | AB017365.1 | - | 1 | 1400 | ||
| BC015915.1 | - | 1 | 1353 | 4549389 | ||
More Information
Set Membership: |
|

FZD7
Genomic Map