siRNA Details
| Gene Symbol: Snai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107439.1 | NP_001100909.1 | 2 | 462 | ||
| XM_006255765.2 | XP_006255827.1 | 2 | 872 | |||
| XM_008772611.1 | XP_008770833.1 | 2 | 3885 | |||
| XM_008772612.1 | XP_008770834.1 | 3 | 5129 | |||

Snai3
Genomic Map