siRNA Details
| Gene Symbol: Zfp276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728603.4 | XP_002728649.1 | 7 | 1725 | ||
| GenBank mRNA | BC079416.1 | - | 7 | 1084 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp276| Gene Symbol: Zfp276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728603.4 | XP_002728649.1 | 7 | 1725 | ||
| GenBank mRNA | BC079416.1 | - | 7 | 1084 | ||