siRNA Details
| Gene Symbol: Map10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277391.1 | NP_001264320.1 | 1 | 872 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map10| Gene Symbol: Map10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277391.1 | NP_001264320.1 | 1 | 872 | ||