siRNA Details
| Gene Symbol: Atxn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772565.1 | XP_008770787.1 | 1 | 1091 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atxn1l| Gene Symbol: Atxn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772565.1 | XP_008770787.1 | 1 | 1091 | ||