siRNA Details
| Gene Symbol: Nrap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107443.2 | NP_001100913.2 | 30 | 3554 | ||
| NM_001113743.1 | NP_001107215.1 | 31 | 3659 | |||
| XM_006231644.2 | XP_006231706.1 | 31 | 3718 | |||
| XM_008760535.1 | XP_008758757.1 | 30 | 3612 | |||

Nrap
Genomic Map