siRNA Details
| Gene Symbol: Nrap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107443.2 | NP_001100913.2 | 34, 35 | 4267 | ||
| NM_001113743.1 | NP_001107215.1 | 35, 36 | 4372 | |||
| XM_006231644.2 | XP_006231706.1 | 35, 36 | 4431 | |||
| XM_008760535.1 | XP_008758757.1 | 34, 35 | 4325 | |||

Nrap
Genomic Map