siRNA Details
| Gene Symbol: Hspa12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107445.1 | NP_001100915.1 | 5 | 1193 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspa12a| Gene Symbol: Hspa12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107445.1 | NP_001100915.1 | 5 | 1193 | ||