siRNA Details
| Gene Symbol: Rab11fip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107447.2 | NP_001100917.2 | 2 | 685 | ||
| GenBank mRNA | BC098050.1 | - | 723 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab11fip2| Gene Symbol: Rab11fip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107447.2 | NP_001100917.2 | 2 | 685 | ||
| GenBank mRNA | BC098050.1 | - | 723 | |||