siRNA Details
| Gene Symbol: GFI1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135031.1 | NP_001128503.1 | 3 | 288 | ||
| NM_004188.6 | NP_004179.3 | 7 | 962 | |||
| XM_006717297.3 | XP_006717360.1 | 3 | 453 | |||
| XM_011519068.2 | XP_011517370.1 | 5 | 853 | |||
| XM_011519069.2 | XP_011517371.1 | 4 | 623 | |||
| XM_011519070.2 | XP_011517372.1 | 5 | 853 | |||
| XM_017015175.1 | XP_016870664.1 | 4 | 583 | |||
| XM_017015176.1 | XP_016870665.1 | 5 | 853 | |||
| GenBank mRNA | AF081946.1 | - | 2 | 137 | ||
| AY428733.1 | - | 3 | 288 | |||
| BC043371.1 | - | 7 | 962 | 5170920 | ||
| BC048254.1 | - | 2 | 168 | |||
| CR536546.1 | - | 2 | 137 | |||

GFI1B
Genomic Map