siRNA Details
| Gene Symbol: Fam204a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107448.2 | NP_001100918.1 | 2, 3 | 404 | ||
| XM_006231673.2 | XP_006231735.1 | 2, 3 | 1220 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam204a| Gene Symbol: Fam204a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107448.2 | NP_001100918.1 | 2, 3 | 404 | ||
| XM_006231673.2 | XP_006231735.1 | 2, 3 | 1220 | |||