siRNA Details
Gene Symbol: Ssx2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175597.3 | NP_783187.1 | 4 | 548 | ||
XM_006233454.2 | XP_006233516.1 | 3 | 361 | |||
XM_006233455.2 | XP_006233517.1 | 3 | 510 | |||
XM_008761498.1 | XP_008759720.1 | 2 | 416 | |||
GenBank mRNA | AF532970.1 | - | 2 | 261 | ||
BC078687.1 | - | 4 | 548 | 7115887 |
Gene Symbol: LOC100909794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749415.3 | XP_003749463.2 | 3 | 368 | ||
XM_006233474.2 | XP_006233536.1 | 3 | 513 | |||
XM_008761540.1 | XP_008759762.1 | 2 | 418 |
