siRNA Details
| Gene Symbol: Pdcd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107452.1 | NP_001100922.1 | 2, 3 | 278 | ||
| XM_006227784.2 | XP_006227846.1 | 2, 3 | 267 | |||
| GenBank mRNA | BC168744.1 | - | 2, 3 | 235 | 6920123 | |
| FQ213638.1 | - | 2, 3 | 199 | |||
| FQ219953.1 | - | 2, 3 | 199 | |||
| FQ219997.1 | - | 2, 3 | 199 | |||
| FQ231349.1 | - | 2, 3 | 199 | |||
| FQ231760.1 | - | 2, 3 | 199 | |||
| FQ234686.1 | - | 2, 3 | 199 | |||

Pdcd6
Genomic Map