siRNA Details
| Gene Symbol: Brd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107453.1 | NP_001100923.1 | 5, 6 | 674 | ||
| XM_006227785.2 | XP_006227847.1 | 5, 6 | 959 | |||
| XM_006227786.2 | XP_006227848.1 | 5, 6 | 959 | |||
| XM_006227787.2 | XP_006227849.1 | 5, 6 | 959 | |||

Brd9
Genomic Map