siRNA Details
| Gene Symbol: Map3k4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107456.1 | NP_001100926.1 | 21 | 4019 | ||
| XM_006227872.2 | XP_006227934.1 | 24 | 4576 | |||
| XM_006227873.2 | XP_006227935.1 | 23 | 4414 | |||
| XM_006227874.2 | XP_006227936.1 | 23 | 4333 | |||

Map3k4
Genomic Map