siRNA Details
| Gene Symbol: Zfp541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748775.3 | XP_003748823.1 | 9, 10 | 3533 | ||
| XM_006228357.2 | XP_006228419.1 | 10, 11 | 3598 | |||
| XM_006228358.2 | XP_006228420.1 | 9, 10 | 3270 | |||
| XM_008759027.1 | XP_008757249.1 | 11, 12 | 3708 | |||
| XM_008759028.1 | XP_008757250.1 | 9, 10 | 3317 | |||
| XM_008759029.1 | XP_008757251.1 | 10, 11 | 3643 | |||

Zfp541
Genomic Map