siRNA Details
| Gene Symbol: Tmem35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001799.1 | NP_001001799.1 | 2 | 716 | ||
| GenBank mRNA | AY540309.1 | - | 2 | 716 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem35| Gene Symbol: Tmem35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001799.1 | NP_001001799.1 | 2 | 716 | ||
| GenBank mRNA | AY540309.1 | - | 2 | 716 | ||