siRNA Details
| Gene Symbol: Tfb1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181474.2 | NP_852139.2 | 3 | 357 | ||
| GenBank mRNA | AY198392.1 | - | 3 | 337 | ||
| BC070907.1 | - | 3 | 357 | 7102927 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tfb1m| Gene Symbol: Tfb1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181474.2 | NP_852139.2 | 3 | 357 | ||
| GenBank mRNA | AY198392.1 | - | 3 | 337 | ||
| BC070907.1 | - | 3 | 357 | 7102927 | ||