siRNA Details
Gene Symbol: Cyp26c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_217935.5 | XP_217935.3 | 5 | 2366 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cyp26c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_217935.5 | XP_217935.3 | 5 | 2366 |