siRNA Details
| Gene Symbol: Chd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107465.1 | NP_001100935.1 | 7 | 1128 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chd1| Gene Symbol: Chd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107465.1 | NP_001100935.1 | 7 | 1128 | ||