siRNA Details
| Gene Symbol: Phactr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214458.1 | NP_999623.1 | 10 | 1538 | ||
| XM_008758610.1 | XP_008756832.1 | 11 | 2695 | |||
| XM_008758611.1 | XP_008756833.1 | 11 | 2468 | |||
| GenBank mRNA | AY500158.1 | - | 10 | 1538 | ||

Phactr2
Genomic Map