siRNA Details
| Gene Symbol: LOC103689982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758837.1 | XP_008757059.1 | 7 | 1239 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103689982| Gene Symbol: LOC103689982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758837.1 | XP_008757059.1 | 7 | 1239 | ||