siRNA Details
| Gene Symbol: Tmc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034104.1 | NP_001029276.1 | 1235 | |||
| GenBank mRNA | BC100657.1 | - | 1235 | 7936370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmc4| Gene Symbol: Tmc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034104.1 | NP_001029276.1 | 1235 | |||
| GenBank mRNA | BC100657.1 | - | 1235 | 7936370 | ||