siRNA Details
| Gene Symbol: Nlrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107474.1 | NP_001100944.1 | 3 | 1510 | ||
| XM_008758920.1 | XP_008757142.1 | 1 | 1382 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp5| Gene Symbol: Nlrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107474.1 | NP_001100944.1 | 3 | 1510 | ||
| XM_008758920.1 | XP_008757142.1 | 1 | 1382 | |||