siRNA Details
Gene Symbol: Nlrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107474.1 | NP_001100944.1 | 9 | 2784 | ||
XM_008758920.1 | XP_008757142.1 | 7 | 2656 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nlrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107474.1 | NP_001100944.1 | 9 | 2784 | ||
XM_008758920.1 | XP_008757142.1 | 7 | 2656 |