siRNA Details
| Gene Symbol: Zfp667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008557.1 | NP_001008557.1 | 5 | 1453 | ||
| XM_006228189.2 | XP_006228251.1 | 5 | 1792 | |||
| GenBank mRNA | AY221750.1 | - | 5 | 1453 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp667| Gene Symbol: Zfp667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008557.1 | NP_001008557.1 | 5 | 1453 | ||
| XM_006228189.2 | XP_006228251.1 | 5 | 1792 | |||
| GenBank mRNA | AY221750.1 | - | 5 | 1453 | ||