siRNA Details
| Gene Symbol: Zfp579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126276.1 | NP_001119748.1 | 2 | 481 | ||
| XM_006228256.2 | XP_006228318.1 | 2 | 952 | |||
| GenBank mRNA | BC161912.1 | - | 2 | 481 | 8369301 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp579| Gene Symbol: Zfp579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126276.1 | NP_001119748.1 | 2 | 481 | ||
| XM_006228256.2 | XP_006228318.1 | 2 | 952 | |||
| GenBank mRNA | BC161912.1 | - | 2 | 481 | 8369301 | |