siRNA Details
Gene Symbol: Tmem150b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107476.1 | NP_001100946.1 | 2 | 392 | ||
XM_006228267.2 | XP_006228329.1 | 3 | 802 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem150b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107476.1 | NP_001100946.1 | 2 | 392 | ||
XM_006228267.2 | XP_006228329.1 | 3 | 802 |