siRNA Details
| Gene Symbol: Ppp1r37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107482.1 | NP_001100952.1 | 7 | 1124 | ||
| GenBank mRNA | BC166755.1 | - | 7 | 934 | 7930156 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r37| Gene Symbol: Ppp1r37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107482.1 | NP_001100952.1 | 7 | 1124 | ||
| GenBank mRNA | BC166755.1 | - | 7 | 934 | 7930156 | |