siRNA Details
| Gene Symbol: Zfp112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107487.1 | NP_001100957.1 | 4 | 2158 | ||
| XM_006228422.2 | XP_006228484.1 | 5 | 2322 | |||
| XM_008758926.1 | XP_008757148.1 | 4 | 2182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp112| Gene Symbol: Zfp112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107487.1 | NP_001100957.1 | 4 | 2158 | ||
| XM_006228422.2 | XP_006228484.1 | 5 | 2322 | |||
| XM_008758926.1 | XP_008757148.1 | 4 | 2182 | |||